2-Fluoro-3-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A116602-10G A116602-250MG A116602-25G A116602-1G
103798-762EA 16 USD
103798-762 103824-626 76781-418 76781-420 76781-422 77311-430
2-Fluoro-3-nitrobenzonitrile 97%
2-Fluoro-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11849938
CAS Number: 1214328-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR