2-Fluoro-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A175561-100G A175561-10G A175561-500G A175561-1G A175561-25G A175561-5G
103807-056EA 5 USD
103807-056 103845-368 77260-068 77260-070 77260-072 77301-382 77301-384 77301-386
2-Fluoro-3-nitrobenzoic acid 98%
2-Fluoro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
Boiling Pt: 347.6 °C (760 mm Hg)
Flash Pt: 198.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797825
CAS Number: 317-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR