2-Fluoro-3-methoxybenzonitrile 98%

Supplier: Ambeed
Warning

A938629-1G A938629-25G A938629-5G A938629-10G A938629-250MG
103841-422EA 73 USD
103841-422 76845-684 76845-686 76845-688 77036-980 77036-982
2-Fluoro-3-methoxybenzonitrile 98%
2-Fluoro-3-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD09839223
CAS Number: 198203-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR