2-Fluoro-3-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A313457-25G A313457-100G A313457-10G A313457-1G A313457-5G
103825-004EA 7 USD
103825-004 103835-916 76838-734 77002-732 77002-734 77306-086 77306-088
2-Fluoro-3-methoxybenzoic acid 97%
2-Fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Boiling Pt: 296.6 °C (760 mm Hg)
Melting Pt: 157…160 °C
Flash Pt: 133.2 °C
Storage Temperature: Ambient
MDL Number: MFCD02179621
CAS Number: 137654-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR