2-Fluoro-m-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-3-methoxyaniline

A103390-100MG A103390-250MG A103390-1G
103796-542EA 23 USD
103796-542 103822-246 77025-272 77025-274 77283-088
2-Fluoro-m-anisidine 98%
2-Fluoro-m-anisidine

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.1400 g/mol
Boiling Pt: 231 °C (760 mm Hg)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11042216
CAS Number: 801282-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR