2-Fluoro-3-iodobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A126287-250MG
A126287-1G
A126287-5G
A126287-10G
103800-790EA
16
USD
103800-790
77026-824
77298-110
77312-996
77666-796
2-Fluoro-3-iodobenzoic acid 95%
2-Fluoro-3-iodobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄FIO₂ Flash Pt: 153.994 °C Storage Temperature: Ambient |
MDL Number:
MFCD07774248 CAS Number: 447464-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...