2-Fluoro-3-formylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoronicotinaldehyde

A204224-1G A204224-25G A204224-5G A204224-100G A204224-10G
103817-534EA 6 USD
103817-534 103848-196 77086-352 77086-354 77086-356 77164-704 77164-706
2-Fluoro-3-formylpyridine 95%
2-Fluoro-3-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄FNO
MW: 125.1 g/mol
Boiling Pt: 220.8 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD03095270
CAS Number: 36404-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR