1,4-Dimethoxy-2-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1,4-dimethoxybenzene

A221173-1G A221173-5G A221173-100G
103818-904EA 9 USD
103818-904 103849-784 77088-380 77088-382 77192-888
1,4-Dimethoxy-2-fluorobenzene 98%
1,4-Dimethoxy-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Boiling Pt: 198 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00042487
CAS Number: 82830-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR