1,4-Diiodo-2-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1,4-diiodobenzene

A153546-10G A153546-250MG A153546-1G A153546-25G A153546-5G
103804-524EA 9 USD
103804-524 103842-698 77190-876 77190-878 77318-362 77318-364 77318-366
1,4-Diiodo-2-fluorobenzene 98%
1,4-Diiodo-2-fluorobenzene

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₆H₃FI₂
MW: 347.9000 g/mol
Boiling Pt: 272.7 °C (760 mm Hg)
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00061107
CAS Number: 147808-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR