2-Ethynylbenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Formylphenylacetylene

A106987-10G A106987-250MG A106987-25G A106987-1G A106987-5G
103797-104EA 19 USD
103797-104 103822-862 77257-486 77257-488 77257-490 77309-730 77309-732
2-Ethynylbenzaldehyde 97%
2-Ethynylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O
MW: 130.143 g/mol
Boiling Pt: 230.3 °C (760 mm Hg)
Melting Pt: 64…67 °C
Flash Pt: 90.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04039985
CAS Number: 38846-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR