2-Ethynylaniline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminophenylacetylene

A349343-1G A349343-25G A349343-5G A349343-10G
103826-016EA 14 USD
103826-016 103837-006 76840-096 76840-098 76840-100 77263-842
2-Ethynylaniline 97%
2-Ethynylaniline

Precaución: for Research Use Only

Formula: C₈H₇N
Boiling Pt: 229.5 °C (760 mm Hg)
Flash Pt: 98.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168862
CAS Number: 52670-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR