2-Ethylphenylboronic acid 97%

Supplier: Ambeed
Warning

A148237-10G A148237-1G A148237-5G
103803-896EA 5 USD
103803-896 103842-012 77190-678 77317-450 77317-452
2-Ethylphenylboronic acid 97%
2-Ethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₂
Boiling Pt: 299.1 °C (760 mm Hg)
Melting Pt: 102.5…107.5 °C
Flash Pt: 134.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02093075
CAS Number: 90002-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR