2-Ethylhexyl stearate 97%

Supplier: Ambeed
Warning

A377025-5G A377025-100G
103837-626EA 5 USD
103837-626 76840-930 77264-174 77264-176
2-Ethylhexyl stearate 97%
2-Ethylhexyl stearate

Precaución: for Research Use Only

Formula: C₂₆H₅₂O₂
Boiling Pt: 431.8 °C (760 mm Hg)
Flash Pt: 219.202 °C
Storage Temperature: Ambient
MDL Number: MFCD00072275
CAS Number: 22047-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR