2-Ethylhexyl 4-aminobenzoate 97%
Supplier: Ambeed
|
Warning
|
A108093-100G
A108093-10G
A108093-250MG
A108093-25G
A108093-1G
A108093-5G
103797-294EA
18
USD
103797-294
103823-068
77233-300
77233-302
77233-304
77233-306
77309-912
77309-914
2-Ethylhexyl 4-aminobenzoate 97%
2-Ethylhexyl 4-aminobenzoate
Precaución: for Research Use Only
Formula:
C₁₅H₂₃NO₂ Boiling Pt: 384.962 °C (760 mm Hg) Flash Pt: 222.018 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13340006 CAS Number: 26218-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...