2-Ethylbenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A140695-100G
A140695-500G
A140695-5G
A140695-1G
A140695-25G
103802-896EA
7
USD
103802-896
76811-750
76811-752
77299-158
77316-004
77316-006
2-Ethylbenzoic acid 98%
2-Ethylbenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀O₂ Boiling Pt: 304.8 °C (760 mm Hg) Melting Pt: 62…66 °C Flash Pt: 138.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045838 CAS Number: 612-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...