2-Ethylbenzaldehyde 97%

Supplier: Ambeed
Warning

A164070-100G A164070-10G A164070-1G A164070-25G A164070-5G
103805-808EA 13 USD
103805-808 103844-074 77259-726 77259-728 77320-206 77320-208 77320-210
2-Ethylbenzaldehyde 97%
2-Ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O
Boiling Pt: 210 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02261766
CAS Number: 22927-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR