2-Ethyl-3-hydroxy-6-methylpyridine succinate 97%

Supplier: Ambeed
Warning

Synonyms: Emoxypine succinate, 2-Ethyl-6-methylpyridin-3-ol succinate

A398276-5G A398276-100G A398276-10G
103827-016EA 5 USD
103827-016 103838-056 76841-490 77601-756 77601-758
2-Ethyl-3-hydroxy-6-methylpyridine succinate 97%
2-Ethyl-3-hydroxy-6-methylpyridine succinate

Precaución: for Research Use Only

Formula: C₈H₁₁NO·C₄H₆O₄
MW: 255.27 g/mol
Boiling Pt: 280.6 °C (760 mm Hg)
Flash Pt: 123.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00445612
CAS Number: 127464-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR