2-Ethyl-6-methyl-3-pyridinol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethyl-6-methylpyridin-3-ol hydrochloride

A101309-100MG A101309-10G A101309-1G
103795-902EA 6 USD
103795-902 103821-588 77257-236 77257-238 77297-052
2-Ethyl-6-methyl-3-pyridinol hydrochloride 97%
2-Ethyl-6-methyl-3-pyridinol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO
MW: 173.64 g/mol
Flash Pt: 123.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00460776
CAS Number: 13258-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR