2-Ethyl-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A137989-10G A137989-1G A137989-25G A137989-5G A137989-100G
103802-544EA 5 USD
103802-544 77095-298 77315-546 77315-548 77315-550 77901-797
2-Ethyl-1H-benzo[d]imidazole 97%
2-Ethyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
Melting Pt: 171…172 °C
Flash Pt: 179.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00022684
CAS Number: 1848-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR