2-Ethoxyphenol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-2-ethoxybenzene, Pyrocatechol monoethyl ether, o-Ethoxyphenol

A145163-100G A145163-500G A145163-25G A145163-5G
103841-578EA 9 USD
103841-578 77137-512 77137-514 77316-850 77316-852
2-Ethoxyphenol 98%
2-Ethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 219.4 °C (760 mm Hg)
Melting Pt: 20…25 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR