2-Ethoxy-2-oxoacetic acid 80%

Supplier: Ambeed
Danger

A260606-100MG A260606-10G A260606-250MG A260606-1G A260606-5G
103812-500EA 58 USD
103812-500 103852-386 76816-002 76816-004 76816-006 77304-798 77304-800
2-Ethoxy-2-oxoacetic acid 80%
2-Ethoxy-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₄H₆O₄
MW: 118.08 g/mol
Flash Pt: 89.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00785870
CAS Number: 617-37-8
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR