2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2-Diphenylphosphino-2'-(N,N-dimethylamino)biphenyl, PhDavePhos, 2-Diphenylphosphino-2'-(N,N-dimethylamino))-1,1'-biphenyl

A186205-100G A186205-250MG A186205-1G A186205-25G A186205-5G A186205-10G
103815-924EA 7 USD
103815-924 103846-500 76998-848 76998-850 77084-152 77084-154 77084-156 77671-630
2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl 98%
2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl

Precaución: for Research Use Only

Formula: C₂₆H₂₄NP
Boiling Pt: 514.6 °C (760 mm Hg)
Melting Pt: 126…128 °C
Flash Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD03094578
CAS Number: 240417-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR