2'-Deoxyguanosine monohydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate
A104006-10G
A104006-1G
A104006-25G
103796-636EA
5
USD
103796-636
103822-348
76809-454
77283-180
77882-893
2'-Deoxyguanosine monohydrate 97%
2'-Deoxyguanosine monohydrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃N₅O₄·H₂O MW: 285.26 g/mol Flash Pt: 438.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150760 CAS Number: 312693-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...