2'-Deoxy-2'-fluorouridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
A100390-100G
A100390-10G
A100390-250MG
A100390-25G
A100390-1G
A100390-5G
103795-412EA
12
USD
103795-412
103821-110
77270-148
77270-150
77270-152
77270-154
77296-664
77296-666
2'-Deoxy-2'-fluorouridine 98%
2'-Deoxy-2'-fluorouridine
Precaución: for Research Use Only
Formula:
C₉H₁₁FN₂O₅ MW: 246.19 g/mol Melting Pt: 149…150 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01317293 CAS Number: 784-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...