2-Cyclopropylethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-Cyclopropylethanamine hydrochloride

A611032-10G A611032-250MG A611032-1G A611032-25G A611032-5G
103840-166EA 95 USD
103840-166 76831-006 76831-008 76844-330 76844-332 76844-334
2-Cyclopropylethylamine hydrochloride 95%
2-Cyclopropylethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁N·ClH
Storage Temperature: Ambient
MDL Number: MFCD14525749
CAS Number: 89381-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR