2-Cyclopropyl-5-iodo-1H-imidazole 97%

Supplier: Ambeed
Warning

A138171-100MG A138171-250MG
103802-572EA 100 USD
103802-572 77246-790 77246-792
2-Cyclopropyl-5-iodo-1H-imidazole 97%
2-Cyclopropyl-5-iodo-1H-imidazole

Precaución: for Research Use Only

Formula: C₂₁H₂₉N₃O₆
Boiling Pt: 407.1±18.0 °C (760 mm Hg)
Flash Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28386135
CAS Number: 761426-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR