2-Cyclopropyl-5-pyridineboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A175857-250MG A175857-100MG A175857-5G
103807-090EA 138 USD
103807-090 77600-422 77871-879 77871-880
2-Cyclopropyl-5-pyridineboronic acid pinacol ester ≥95%
2-Cyclopropyl-5-pyridineboronic acid pinacol ester

Precaución: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₁₄H₂₂BNO₃
Boiling Pt: 354.1 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09952039
CAS Number: 893567-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR