2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A137854-5G
A137854-10G
103802-520EA
8
USD
103802-520
77315-506
77611-254
2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carbaldehyde 95%
2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₉H₁₄FNO Boiling Pt: 453.9 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09032925 CAS Number: 121660-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...