2-Cyclopropyl-3-[(diphenylphosphinyl)methyl]-4-(4-fluorophenyl)quinolin 97%

Supplier: Ambeed
Warning

A143174-100MG A143174-250MG
103803-248EA 51 USD
103803-248 76811-856 76811-858
2-Cyclopropyl-3-[(diphenylphosphinyl)methyl]-4-(4-fluorophenyl)quinolin 97%
2-Cyclopropyl-3-[(diphenylphosphinyl)methyl]-4-(4-fluorophenyl)quinolin

Precaución: for Research Use Only

Formula: C₃₁H₂₅FNOP
MW: 477.5 g/mol
Boiling Pt: 644.3 °C (760 mm Hg)
Flash Pt: 343.4 °C
Storage Temperature: Ambient
MDL Number: MFCD12828197
CAS Number: 146578-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR