Cyclohexylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Cyclohexylboronic acid Pinacol ester, 2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohexane

A237614-100G A237614-1G A237614-25G A237614-5G A237614-10G
103811-164EA 5 USD
103811-164 103850-944 76722-352 77090-034 77090-036 77090-038 77568-298
Cyclohexylboronic acid pinacol ester 98%
Cyclohexylboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₂H₂₃BO₂
Boiling Pt: 244.7 °C (760 mm Hg)
Flash Pt: 101.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038749
CAS Number: 87100-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR