2-Cyano-N-methyl-acetamide 95%

Supplier: Ambeed
Warning

A130923-100G A130923-10G A130923-1G A130923-25G A130923-5G
103801-492EA 6 USD
103801-492 77161-862 77161-864 77298-434 77313-968 77313-970
2-Cyano-N-methyl-acetamide 95%
2-Cyano-N-methyl-acetamide

Precaución: for Research Use Only

Formula: C₄H₆N₂O
MW: 98.1000 g/mol
Melting Pt: 82 °C
Flash Pt: 156.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00090119
CAS Number: 6330-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR