2-Cyano-N,N-diethylacetamide 97%

Supplier: Ambeed
Warning

A124607-100G A124607-25G A124607-1G A124607-5G
103800-556EA 5 USD
103800-556 77108-762 77108-764 77312-692 77312-694
2-Cyano-N,N-diethylacetamide 97%
2-Cyano-N,N-diethylacetamide

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O
MW: 140.19 g/mol
Boiling Pt: 254.1 °C (760 mm Hg)
Flash Pt: 107.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01332383
CAS Number: 26391-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR