2-Cyanoisonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-4-pyridinecarboxylic acid

A154016-10G A154016-250MG A154016-1G A154016-5G A154016-25G
103804-594EA 12 USD
103804-594 103842-774 77259-380 77259-382 77318-458 77318-460 77748-060
2-Cyanoisonicotinic acid 97%
2-Cyanoisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MW: 148.1200 g/mol
Boiling Pt: 478.4 °C (760 mm Hg)
Flash Pt: 243.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03411729
CAS Number: 161233-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR