2-Cyanobenzaldehyde 97%

Supplier: Ambeed
Danger

Synonyms: 2-Formylbenzonitrile

A132497-100G A132497-10G A132497-500G A132497-1G A132497-25G A132497-5G
103801-746EA 5 USD
103801-746 76870-800 76870-802 76870-804 77298-562 77314-324 77314-326
2-Cyanobenzaldehyde 97%
2-Cyanobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅NO
MW: 131.13 g/mol
Melting Pt: 103…105 °C
Flash Pt: 126.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017503
CAS Number: 7468-67-9
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR