5-Iodopyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-5-iodopyridine

A112766-250MG A112766-5G
103798-098EA 69 USD
103798-098 103823-936 76970-948 77297-210
5-Iodopyridine-2-carbonitrile 97%
5-Iodopyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃IN₂
MW: 230.0100 g/mol
Boiling Pt: 314.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10699132
CAS Number: 41960-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR