5-Fluoropyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-pyridinecarbonitrile, 2-Cyano-5-fluoropyridine

A119592-100G A119592-25G A119592-1G A119592-5G A119592-10G
103799-792EA 5 USD
103799-792 76781-644 76781-646 77297-700 77311-910 77592-328
5-Fluoropyridine-2-carbonitrile 98%
5-Fluoropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃FN₂
MW: 122.1 g/mol
Melting Pt: 35…41 °C
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08741371
CAS Number: 327056-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR