2-Cyano-5-fluorobenzyl bromide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Bromomethyl-4-fluorobenzonitrile
A606966-25G
A606966-100G
A606966-10G
A606966-1G
103840-146EA
20
USD
103840-146
76844-312
76980-540
76980-542
77307-934
2-Cyano-5-fluorobenzyl bromide 98%
2-Cyano-5-fluorobenzyl bromide
Precaución: For research use only
Formula:
C₈H₅BrFN Boiling Pt: 264.5 °C (760 mm Hg) Flash Pt: 113.746 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08059542 CAS Number: 421552-12-7 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...