2-Cyano-4-methylpent-2-enoic acid 97%

Supplier: Ambeed
Warning

A191057-5G A191057-250MG
103816-342EA 224 USD
103816-342 77236-886 77593-216
2-Cyano-4-methylpent-2-enoic acid 97%
2-Cyano-4-methylpent-2-enoic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 283.1 °C (760 mm Hg)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD22377852
CAS Number: 760-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR