2-Chlorothiazolo[4,5-b]pyrazine 95%

Supplier: Ambeed
Warning

A191179-100MG A191179-250MG A191179-1G
103816-350EA 416 USD
103816-350 103846-952 77139-040 77139-042 77302-248
2-Chlorothiazolo[4,5-b]pyrazine 95%
2-Chlorothiazolo[4,5-b]pyrazine

Precaución: for Research Use Only

Formula: C₅H₂ClN₃S
MW: 171.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29071823
CAS Number: 1190927-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR