2-Chloroquinoline-6-carboxylic acid 96%

Supplier: Ambeed
Warning

A226254-100MG A226254-250MG A226254-5G
103819-234EA 119 USD
103819-234 103850-134 76786-128 76786-130 77088-862
2-Chloroquinoline-6-carboxylic acid 96%
2-Chloroquinoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
Boiling Pt: 376.1 °C (760 mm Hg)
Flash Pt: 181.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09730432
CAS Number: 849996-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR