2-Chloroquinoline-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A133668-100G A133668-10G A133668-1G A133668-25G A133668-5G
103801-898EA 8 USD
103801-898 76797-958 76797-960 77314-572 77314-574 77314-576
2-Chloroquinoline-3-carbaldehyde 95%
2-Chloroquinoline-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO
Melting Pt: 148…150 °C
Flash Pt: 156.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00130079
CAS Number: 73568-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR