2-Chloroquinoline 98%

Supplier: Ambeed
Warning

A128551-100G A128551-10G A128551-500G A128551-1G A128551-25G A128551-5G
103801-152EA 5 USD
103801-152 77258-366 77258-368 77258-370 77313-478 77313-480 77313-482
2-Chloroquinoline 98%
2-Chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₆ClN
Boiling Pt: 267.3 °C (760 mm Hg)
Melting Pt: 34…37 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006741
CAS Number: 612-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR