2-Chloroquinolin-4-amine 98%

Supplier: Ambeed
Warning

A124299-10G A124299-250MG A124299-50MG A124299-5G A124299-1G
103800-512EA 53 USD
103800-512 77285-040 77285-042 77285-044 77297-982 77312-648
2-Chloroquinolin-4-amine 98%
2-Chloroquinolin-4-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MW: 178.62 g/mol
Boiling Pt: 365.8 °C (760 mm Hg)
Melting Pt: 250 °C
Flash Pt: 175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08705643
CAS Number: 80947-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR