2-Chloropyrimidine-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A170664-1G
A170664-25G
A170664-5G
A170664-100G
A170664-10G
A170664-250MG
103806-526EA
11
USD
103806-526
103844-800
77081-910
77081-912
77081-914
77163-556
77163-558
77163-560
2-Chloropyrimidine-5-carboxylic acid 97%
2-Chloropyrimidine-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₃ClN₂O₂ Boiling Pt: 411.4 °C (760 mm Hg) Flash Pt: 202.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06739036 CAS Number: 374068-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...