2-Chloropyrimidine-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A212951-100G
A212951-10G
A212951-250MG
A212951-25G
A212951-1G
A212951-5G
103818-252EA
11
USD
103818-252
103848-950
76986-784
76986-786
76986-788
76986-790
77087-428
77087-430
2-Chloropyrimidine-4-carboxylic acid 98%
2-Chloropyrimidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₃ClN₂O₂ MW: 158.5400 g/mol Boiling Pt: 411.4 °C (760 mm Hg) Melting Pt: 197…201 °C Flash Pt: 118.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06739041 CAS Number: 149849-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...