2-Chloropyrimidine-4-carbonitrile 98%

Supplier: Ambeed
Warning

A182805-10G A182805-1G A182805-25G A182805-5G
103815-572EA 25 USD
103815-572 103846-130 76769-610 77083-696 77083-698 77083-700
2-Chloropyrimidine-4-carbonitrile 98%
2-Chloropyrimidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂ClN₃
MW: 139.5400 g/mol
Boiling Pt: 331.3 °C (760 mm Hg)
Melting Pt: 53…55 °C
Flash Pt: 188.372 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234989
CAS Number: 75833-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR