2-Chloropyrimidine-4,5-diamine 97%

Supplier: Ambeed
Warning

A159308-100MG A159308-1G
103805-244EA 23 USD
103805-244 103843-462 77011-136 77300-398
2-Chloropyrimidine-4,5-diamine 97%
2-Chloropyrimidine-4,5-diamine

Precaución: for Research Use Only

Formula: C₄H₅ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD00047355
CAS Number: 14631-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR