2-Chloro-4-hydroxypyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloropyrimidin-4-ol

A107887-250MG A107887-10G
103797-256EA 24 USD
103797-256 103823-028 76970-540 77565-890
2-Chloro-4-hydroxypyrimidine 97%
2-Chloro-4-hydroxypyrimidine

Precaución: For research use only.

Formula: C₄H₃ClN₂O
MW: 130.532 g/mol
Boiling Pt: 328.1 °C (760 mm Hg)
Flash Pt: 152.3 °C
Storage Temperature: Freezer
MDL Number: MFCD09038284
CAS Number: 55873-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR