2-Chloro-pyrido[2,3-b]pyrazine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloropyrido[2,3-b]pyrazine

A244901-100MG A244901-250MG
103811-610EA 62 USD
103811-610 77013-856 77013-858
2-Chloro-pyrido[2,3-b]pyrazine 95%
2-Chloro-pyrido[2,3-b]pyrazine

Precaución: for Research Use Only

Formula: C₇H₄ClN₃
MW: 165.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738667
CAS Number: 70838-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR