2-Chloropyridine-5-acetonitrile 98%

Supplier: Ambeed
Danger

A529103-1G A529103-25G A529103-5G A529103-10G
103828-556EA 5 USD
103828-556 103839-664 76843-678 76843-680 76843-682 77103-302
2-Chloropyridine-5-acetonitrile 98%
2-Chloropyridine-5-acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
Boiling Pt: 294.1 °C (760 mm Hg)
Melting Pt: 49…54 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112495
CAS Number: 39891-09-3
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR